siRNA Details
Gene Symbol: Cep41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ278891.1 | - | 1, 2 | 65 | ||
AK019270.1 | - | 1, 2 | 54 | |||
AK132495.1 | - | 1, 2 | 54 | |||
AK161461.1 | - | 1, 2 | 39 | |||
AK166974.1 | - | 1, 2 | 46 | |||
AK170213.1 | - | 1, 2 | 78 | |||
BC031892.1 | - | 1, 2 | 35 | 5365552 | ||
BY712226.1 | - | 1, 2 | 54 | |||
BY736279.1 | - | 1, 2 | 46 |
