siRNA Details
Gene Symbol: Cep41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ278891.1 | - | 7 | 592 | ||
AK006059.1 | - | 7 | 580 | |||
AK132495.1 | - | 7 | 581 | |||
AK161461.1 | - | 7 | 566 | |||
AK166974.1 | - | 8 | 709 | |||
AK170213.1 | - | 7 | 605 | |||
BC031892.1 | - | 7 | 562 | 5365552 |
