siRNA Details
Gene Symbol: Cep41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ278891.1 | - | 10, 11 | 1018 | ||
AK006059.1 | - | 10, 11 | 1006 | |||
AK132495.1 | - | 10, 11 | 1007 | |||
AK161461.1 | - | 10, 11 | 992 | |||
AK166974.1 | - | 11, 12 | 1135 | |||
AK170213.1 | - | 10, 11 | 1031 | |||
BC031892.1 | - | 10, 11 | 988 | 5365552 |
