siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163641.1 | NP_001157113.1 | 21, 22 | 4002 | ||
NM_001163642.1 | NP_001157114.1 | 21, 22 | 3961 | |||
NM_018877.3 | NP_061365.3 | 21, 22 | 3964 | |||
GenBank mRNA | AF091628.1 | - | 21, 22 | 3915 | ||
AK088590.1 | - | 14, 15 | 2953 | |||
AK122198.1 | - | 17, 18 | 4467 | |||
AK132202.1 | - | 21, 22 | 3937 | |||
BC007176.1 | - | 8, 9 | 3094 | |||
BC079537.1 | - | 20, 21 | 4994 | 6410215 |
