siRNA Details
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163641.1 | NP_001157113.1 | 20 | 3866 | ||
NM_001163642.1 | NP_001157114.1 | 20 | 3825 | |||
NM_018877.3 | NP_061365.3 | 20 | 3828 | |||
GenBank mRNA | AF091628.1 | - | 20 | 3779 | ||
AK088590.1 | - | 13 | 2817 | |||
AK122198.1 | - | 16 | 4331 | |||
AK132202.1 | - | 20 | 3801 | |||
BC007176.1 | - | 7 | 2958 | |||
BC060173.1 | - | 20 | 3820 | |||
BC079537.1 | - | 19 | 4858 | 6410215 |
