siRNA Details
Gene Symbol: Zkscan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251833.1 | NP_001238762.1 | 4 | 862 | ||
NM_139141.4 | NP_631880.2 | 6 | 1078 | |||
GenBank mRNA | AK032718.1 | - | 6 | 1034 | ||
AK134299.1 | - | 6 | 1063 | |||
BC050152.1 | - | 6 | 1014 | |||
BC145448.1 | - | 3 | 696 | 9054006 | ||
BC150775.1 | - | 3 | 696 | 9087686 |
