siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 7 | 1963 | ||
GenBank mRNA | AF330197.1 | - | 7 | 1959 | ||
AK035242.1 | - | 1 | 283 | |||
AK036351.1 | - | 6 | 1698 | |||
AK048110.1 | - | 6 | 1729 | |||
AK137148.1 | - | 5 | 1301 | |||
AK169999.1 | - | 6 | 2072 | |||
BC011463.1 | - | 1 | 395 | |||
BC069835.1 | - | 7 | 1819 | 30094289 |
