siRNA Details
Gene Symbol: Trim16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053169.2 | NP_444399.2 | 5 | 1291 | ||
GenBank mRNA | AF449496.1 | - | 5 | 1042 | ||
AK053139.1 | - | 5 | 1105 | |||
AK078902.1 | - | 3 | 627 | |||
BC052821.1 | - | 6 | 993 | 30057830 |
