siRNA Details
Gene Symbol: Ddx50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053183.2 | NP_444413.1 | 12 | 1774 | ||
GenBank mRNA | AF334104.1 | - | 12 | 1775 | ||
AK030239.1 | - | 13 | 1846 | |||
AK041281.1 | - | 12 | 1673 | |||
AK162347.1 | - | 3 | 2149 | |||
BC049906.1 | - | 12 | 1775 | |||
BC055440.1 | - | 12 | 1769 | |||
BC075714.1 | - | 12 | 1727 | |||
BC090996.1 | - | 12 | 1794 | 6757408 |
