siRNA Details
Gene Symbol: Prg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110146.1 | NP_001103616.1 | 7 | 546 | ||
NM_021400.3 | NP_067375.3 | 8 | 2940 | |||
GenBank mRNA | AB034730.1 | - | 9 | 2439 | ||
AK132597.1 | - | 2 | 686 | |||
AK149469.1 | - | 7 | 547 | |||
BC130021.1 | - | 6 | 510 |
