siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 17 | 1643 | ||
GenBank mRNA | AK004345.1 | - | 7 | 524 | ||
AK034867.1 | - | 16 | 1504 | |||
AK161445.1 | - | 15 | 1482 | |||
AK182433.1 | - | 1 | 50 | |||
AK188028.1 | - | 1 | 50 | |||
BC012436.1 | - | 15 | 1171 | |||
BC018294.1 | - | 17 | 1593 | 4021243 | ||
DV648379.1 | - | 9 | 702 | |||
DV653632.1 | - | 5 | 387 |
