siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 12 | 1216 | ||
GenBank mRNA | AK004345.1 | - | 2 | 97 | ||
AK034867.1 | - | 11 | 1077 | |||
AK161445.1 | - | 10 | 1055 | |||
AK177131.1 | - | 2 | 91 | |||
AK178753.1 | - | 2 | 91 | |||
AK214695.1 | - | 4 | 269 | |||
BC012436.1 | - | 10 | 744 | |||
BC018294.1 | - | 12 | 1166 | 4021243 | ||
DV648379.1 | - | 4 | 275 |
