siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026849.2 | NP_081125.2 | 19 | 2023 | ||
GenBank mRNA | AK004345.1 | - | 8 | 748 | ||
AK034867.1 | - | 18 | 1884 | |||
BC012436.1 | - | 17 | 1551 | |||
BC018294.1 | - | 19 | 1973 | 4021243 | ||
DV653288.1 | - | 2 | 164 |
