siRNA Details
Gene Symbol: Kmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133809.1 | NP_598570.1 | 13 | 1207 | ||
GenBank mRNA | AK133074.1 | - | 1129 | |||
AK171812.1 | - | 13 | 1335 | |||
BC014683.1 | - | 13 | 1207 | 4210278 | ||
BC024455.1 | - | 2 | 159 |
