siRNA Details
Gene Symbol: Cyp2j11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004141.2 | NP_001004141.2 | 3 | 537 | ||
GenBank mRNA | AK143983.1 | - | 3 | 519 | ||
BC080831.1 | - | 3 | 513 | 30469795 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp2j11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004141.2 | NP_001004141.2 | 3 | 537 | ||
GenBank mRNA | AK143983.1 | - | 3 | 519 | ||
BC080831.1 | - | 3 | 513 | 30469795 |