siRNA Details
Gene Symbol: Rdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290411.1 | NP_001277340.1 | 4 | 532 | ||
NM_175372.4 | NP_780581.1 | 4 | 499 | |||
GenBank mRNA | AK028434.1 | - | 4 | 492 | ||
BC082583.1 | - | 4 | 451 | 30613493 |
