siRNA Details
Gene Symbol: Add2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271857.1 | NP_001258786.1 | 10 | 1486 | ||
NM_001271858.1 | NP_001258787.1 | 10 | 1483 | |||
NM_001271859.1 | NP_001258788.1 | 10 | 1284 | |||
NM_001271860.1 | NP_001258789.1 | 10 | 1486 | |||
NM_001271861.1 | NP_001258790.1 | 10 | 1287 | |||
NM_013458.5 | NP_038486.2 | 10 | 1287 | |||
GenBank mRNA | AF100422.1 | - | 9 | 1162 | ||
AF100423.1 | - | 9 | 1162 | |||
AF189769.1 | - | 8 | 1084 | |||
AK014496.1 | - | 10 | 1252 | |||
AK028425.1 | - | 10 | 1258 | |||
AK029488.1 | - | 10 | 1437 | |||
AK147544.1 | - | 10 | 1359 | |||
AK156954.1 | - | 10 | 1276 | |||
BC046783.1 | - | 10 | 1420 | 6401200 | ||
BC053032.1 | - | 10 | 1420 | 6401200 |
