siRNA Details
Gene Symbol: Scgb3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289643.1 | NP_001276572.1 | 1 | 154 | ||
NM_001289644.1 | NP_001276573.1 | 1 | 154 | |||
GenBank mRNA | AF274959.1 | - | 1 | 128 | ||
AF274960.1 | - | 1 | 128 | |||
AF274961.1 | - | 1 | 128 | |||
AF439546.1 | - | 1 | 120 | |||
AK136373.1 | - | 1 | 132 | |||
AK136396.1 | - | 1 | 131 | |||
BC061046.1 | - | 128 | 30301032 | |||
BY120182.1 | - | 1 | 155 |
