siRNA Details
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301250.1 | NP_001288179.1 | 12 | 1713 | ||
NM_007602.4 | NP_031628.1 | 12 | 1877 | |||
GenBank mRNA | AK139476.1 | - | 1 | 26 | ||
AK155179.1 | - | 12 | 1855 | |||
AK164293.1 | - | 12 | 1787 | |||
AK170034.1 | - | 12 | 1814 | |||
BC014767.1 | - | 12 | 1705 | 4458488 | ||
U85020.1 | - | 12 | 1796 | |||
Y10656.1 | - | 12 | 1809 |
