siRNA Details
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007622.3 | NP_031648.1 | 3 | 510 | ||
GenBank mRNA | AK014215.1 | - | 3 | 486 | ||
AK017608.1 | - | 3 | 399 | |||
AK157710.1 | - | 3 | 512 | |||
BC055290.1 | - | 3 | 453 | 6313666 | ||
X56690.1 | - | 2 | 255 |
