siRNA Details
Gene Symbol: Col4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009932.3 | NP_034062.3 | 46 | 4861 | ||
GenBank mRNA | AY375463.1 | - | 1 | 90 | ||
AY502946.1 | - | 1 | 90 | |||
BC013560.1 | - | 4 | 518 | |||
BC138040.1 | - | 45 | 4573 | 8861059 | ||
J04695.1 | - | 46 | 4666 | |||
M15833.1 | - | 1 | 90 | |||
X04647.1 | - | 14 | 1623 |
