siRNA Details
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037726.1 | NP_001032815.1 | 5 | 631 | ||
NM_009952.2 | NP_034082.1 | 7 | 795 | |||
NM_133828.2 | NP_598589.2 | 6 | 753 | |||
GenBank mRNA | AF448507.1 | - | 7 | 671 | ||
AF448508.1 | - | 1323 | ||||
AK042595.1 | - | 6 | 726 | |||
AK044992.1 | - | 6 | 568 | |||
AK044993.1 | - | 6 | 568 | |||
AK079223.1 | - | 6 | 749 | |||
BB831364.1 | - | 2 | 122 | |||
BC021649.1 | - | 6 | 686 | 4017170 | ||
M95106.1 | - | 7 | 705 | |||
X92497.1 | - | 5 | 608 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031017, NM_134443, XM_008767138, XM_006245067, XM_006245069, and others.
Gene Ontology Categories:
