siRNA Details
Gene Symbol: Crkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007764.5 | NP_031790.2 | 2 | 1106 | ||
GenBank mRNA | AK048641.1 | - | 2 | 1104 | ||
AK052315.1 | - | 2 | 1106 | |||
AK163981.1 | - | 2 | 1078 | |||
BC131984.1 | - | 2 | 873 | 40130261 | ||
BC131986.1 | - | 2 | 873 | 40130263 | ||
X90648.1 | - | 2 | 945 |
