siRNA Details
Gene Symbol: Zfp358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080461.2 | NP_536709.2 | 2 | 502 | ||
GenBank mRNA | AF397208.1 | - | 2 | 467 | ||
AK155446.1 | - | 2 | 504 | |||
BC035489.1 | - | 1 | 58 | |||
BC040067.1 | - | 1 | 176 | 4008695 | ||
BC055855.1 | - | 1 | 703 | |||
BY749799.1 | - | 2 | 470 |
