siRNA Details
Gene Symbol: Fxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008044.2 | NP_032070.1 | 2, 3 | 268 | ||
GenBank mRNA | AK148147.1 | - | 2, 3 | 290 | ||
AK160458.1 | - | 2, 3 | 278 | |||
BC016644.1 | - | 1, 2 | 981 | |||
BC038010.1 | - | 1, 2 | 992 | |||
BC058533.1 | - | 2, 3 | 250 | |||
BC125337.1 | - | 2, 3 | 266 | 40129852 | ||
BC132337.1 | - | 2, 3 | 266 | 40130614 | ||
U95736.1 | - | 2, 3 | 268 |
Gene Symbol: Gm5037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF223569.1 | - | 1 | 141 |
