siRNA Details
Gene Symbol: Hipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136065.2 | NP_001129537.1 | 7 | 2037 | ||
NM_001294143.1 | NP_001281072.1 | 7 | 2113 | |||
NM_001294144.1 | NP_001281073.1 | 7 | 2113 | |||
NM_010433.2 | NP_034563.2 | 7 | 2113 | |||
GenBank mRNA | AF077659.1 | - | 7 | 1876 | ||
AF170301.1 | - | 7 | 1863 | |||
AF170302.1 | - | 7 | 1863 | |||
AF208292.1 | - | 7 | 1950 | |||
AF273680.1 | - | 7 | 1834 | |||
AF333791.1 | - | 7 | 1740 | |||
AF333792.1 | - | 7 | 1740 | |||
AK019821.1 | - | 3 | 329 | |||
BC031904.1 | - | 7 | 1846 | 5368577 |
