siRNA Details
Gene Symbol: Il2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008367.3 | NP_032393.3 | 2 | 371 | ||
GenBank mRNA | AK037369.1 | - | 2 | 357 | ||
AK088594.1 | - | 2 | 313 | |||
AK171665.1 | - | 2 | 308 | |||
BC114437.1 | - | 2 | 277 | 40054387 | ||
K02891.1 | - | 2 | 254 | |||
M30856.1 | - | 2 | 301 | |||
M54934.1 | - | 2 | 174 |
