siRNA Details
Gene Symbol: Klf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008452.2 | NP_032478.2 | 2 | 868 | ||
GenBank mRNA | AK131962.1 | - | 2 | 869 | ||
BC138737.1 | - | 2 | 876 | 8861759 | ||
BC138738.1 | - | 2 | 876 | 8861760 | ||
U25096.1 | - | 2 | 897 |
