siRNA Details
Gene Symbol: Olfr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146914.2 | NP_667125.1 | 2 | 138 | ||
GenBank mRNA | BC104074.1 | - | 1 | 23 | ||
BC104075.1 | - | 1 | 33 | 40041745 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
