siRNA Details
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138676.2 | NP_619617.2 | 8 | 668 | ||
GenBank mRNA | AF246218.1 | - | 8 | 674 | ||
AK047434.1 | - | 6 | 799 | |||
AK154779.1 | - | 8 | 657 | |||
AK160641.1 | - | 7 | 740 | |||
AK190932.1 | - | 2 | 193 | |||
BC061477.1 | - | 8 | 621 | 6336438 |
