siRNA Details
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147201.2 | NP_671734.1 | 2, 3 | 315 | ||
GenBank mRNA | AF302138.1 | - | 2, 3 | 314 | ||
AK046142.1 | - | 2, 3 | 309 | |||
AK077440.1 | - | 2, 3 | 351 | |||
AK170501.1 | - | 2, 3 | 331 | |||
AK178908.1 | - | 2, 3 | 156 | |||
AK191367.1 | - | 3, 4 | 195 | |||
AK217229.1 | - | 1, 2 | 242 | |||
BC004756.1 | - | 2, 3 | 285 | 3154089 | ||
BC018463.1 | - | 2, 3 | 289 | 3598438 |
