siRNA Details
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104616.1 | NP_001098086.1 | 9 | 1012 | ||
NM_001104617.1 | NP_001098087.1 | 9 | 1418 | |||
NM_009041.3 | NP_033067.2 | 9 | 1418 | |||
GenBank mRNA | AK016711.1 | - | 9 | 1088 | ||
AK088292.1 | - | 9 | 1125 | |||
AK146423.1 | - | 9 | 1076 | |||
AK156370.1 | - | 9 | 1104 | |||
AK162204.1 | - | 9 | 990 | |||
AK168449.1 | - | 9 | 1089 | |||
AK206105.1 | - | 3 | 330 | |||
BC002041.1 | - | 9 | 1063 | |||
BC026786.1 | - | 9 | 1091 | |||
BC034438.1 | - | 9 | 1244 | |||
BC053417.1 | - | 9 | 1092 | 6707873 | ||
X60672.1 | - | 9 | 1031 |
