siRNA Details
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035228.2 | NP_001030305.1 | 3, 4 | 412 | ||
NM_011375.3 | NP_035505.2 | 3, 4 | 624 | |||
GenBank mRNA | AB013302.1 | - | 2, 3 | 163 | ||
AB018048.1 | - | 3, 4 | 352 | |||
AF119416.1 | - | 3, 4 | 402 | |||
AK012961.1 | - | 3, 4 | 393 | |||
AK158594.1 | - | 3, 4 | 446 | |||
AK159000.1 | - | 3, 4 | 401 | |||
AK165726.1 | - | 3, 4 | 414 | |||
BC138557.1 | - | 3, 4 | 329 | 8861577 | ||
BC138559.1 | - | 3, 4 | 327 | 8861579 | ||
Y15003.1 | - | 3, 4 | 385 |
