siRNA Details
Gene Symbol: Rbms3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172121.1 | NP_001165592.1 | 11 | 1159 | ||
NM_001172122.1 | NP_001165593.1 | 11 | 1682 | |||
NM_001172123.1 | NP_001165594.1 | 11 | 1682 | |||
NM_001172124.1 | NP_001165595.1 | 11 | 1682 | |||
NM_001172126.1 | NP_001165597.1 | 11 | 1682 | |||
NM_178660.4 | NP_848775.2 | 10 | 1631 | |||
GenBank mRNA | AK030478.1 | - | 10 | 1291 | ||
AK030609.1 | - | 11 | 1316 | |||
AK032938.1 | - | 10 | 1628 | |||
AK033005.1 | - | 11 | 1325 | |||
AK041098.1 | - | 11 | 1684 | |||
AK046197.1 | - | 11 | 1160 | |||
AK052196.1 | - | 11 | 1663 | |||
BC033604.1 | - | 6 | 486 | |||
BC108402.1 | - | 10 | 1605 | 6516420 | ||
BC117841.1 | - | 10 | 1071 | 40093225 |
