siRNA Details
Gene Symbol: Iglon5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164518.1 | NP_001157990.1 | 5 | 582 | ||
GenBank mRNA | BC116421.1 | - | 2 | 166 | ||
BC116422.1 | - | 2 | 166 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Iglon5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164518.1 | NP_001157990.1 | 5 | 582 | ||
GenBank mRNA | BC116421.1 | - | 2 | 166 | ||
BC116422.1 | - | 2 | 166 |