siRNA Details
Gene Symbol: Adgrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175642.4 | NP_783573.4 | 25, 26 | 3724 | ||
GenBank mRNA | AK044160.1 | - | 9, 10 | 1483 | ||
AK044248.1 | - | 10, 11 | 1158 | |||
AK081421.1 | - | 4, 5 | 1066 | |||
AY168406.1 | - | 24, 25 | 3463 | |||
BC032251.1 | - | 18, 19 | 1941 | |||
BC062105.1 | - | 9, 10 | 900 | |||
BC079670.1 | - | 23, 24 | 2743 | |||
BC099951.1 | - | 25, 26 | 3722 | 30098919 |
