siRNA Details
Gene Symbol: Lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033250.4 | NP_001028422.2 | 4 | 312 | ||
GenBank mRNA | AK016010.1 | - | 5 | 457 | ||
AK144269.1 | - | 390 | ||||
BC115462.1 | - | 4 | 293 | 40100287 | ||
BC115463.1 | - | 4 | 293 | 40100292 |
