siRNA Details
Gene Symbol: Lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033250.4 | NP_001028422.2 | 2 | 156 | ||
GenBank mRNA | AK016010.1 | - | 2 | 161 | ||
AK144269.1 | - | 163 | ||||
BC048759.1 | - | 2 | 120 | |||
BC115462.1 | - | 2 | 137 | 40100287 | ||
BC115463.1 | - | 2 | 137 | 40100292 |
