siRNA Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159750.1 | NP_001153222.1 | 5 | 552 | ||
NM_001159751.1 | NP_001153223.1 | 5 | 448 | |||
NM_011541.4 | NP_035671.1 | 5 | 555 | |||
GenBank mRNA | AK131928.1 | - | 6 | 739 | ||
AK136035.1 | - | 5 | 448 | |||
AK143593.1 | - | 5 | 413 | |||
BC006022.1 | - | 5 | 430 | 3590733 | ||
BC061490.1 | - | 5 | 447 | 2582185 | ||
BC083127.1 | - | 5 | 418 | 6390364 | ||
CD545750.1 | - | 5 | 436 | |||
M18209.1 | - | 4 | 351 | |||
M18210.1 | - | 5 | 409 |
