siRNA Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159750.1 | NP_001153222.1 | 3 | 391 | ||
NM_001159751.1 | NP_001153223.1 | 3 | 284 | |||
NM_011541.4 | NP_035671.1 | 3 | 391 | |||
GenBank mRNA | AK131928.1 | - | 3 | 575 | ||
AK136035.1 | - | 3 | 284 | |||
AK143593.1 | - | 3 | 249 | |||
BC006022.1 | - | 3 | 266 | 3590733 | ||
BC061490.1 | - | 3 | 283 | 2582185 | ||
BC083127.1 | - | 3 | 254 | 6390364 | ||
CD545750.1 | - | 3 | 275 | |||
CJ059479.1 | - | 3 | 392 | |||
M18209.1 | - | 2 | 187 | |||
M18210.1 | - | 3 | 245 |
