siRNA Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159750.1 | NP_001153222.1 | 5 | 578 | ||
NM_001159751.1 | NP_001153223.1 | 5 | 474 | |||
NM_011541.4 | NP_035671.1 | 5 | 581 | |||
GenBank mRNA | AK131928.1 | - | 6 | 765 | ||
AK136035.1 | - | 5 | 474 | |||
AK143593.1 | - | 5 | 439 | |||
BC006022.1 | - | 5 | 456 | 3590733 | ||
BC061490.1 | - | 5 | 473 | 2582185 | ||
BC083127.1 | - | 5 | 444 | 6390364 | ||
CD545750.1 | - | 5 | 462 | |||
M18209.1 | - | 4 | 377 | |||
M18210.1 | - | 5 | 435 |
