siRNA Details
Gene Symbol: Fcgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122603.1 | NP_001116075.1 | 16 | 6670 | ||
GenBank mRNA | AK040441.1 | - | 6 | 3374 | ||
BC024520.1 | - | 1 | 332 | |||
BC026653.1 | - | 2 | 875 | |||
BC029250.1 | - | 4 | 1693 | |||
BC030871.1 | - | 5 | 2519 | |||
BC036158.1 | - | 2 | 712 | |||
BC039286.1 | - | 7 | 3096 | |||
BC055475.1 | - | 2 | 383 | |||
BC069956.1 | - | 2 | 678 | |||
BC092067.1 | - | 2 | 749 |
