siRNA Details
Gene Symbol: Fcgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122603.1 | NP_001116075.1 | 15 | 6230 | ||
GenBank mRNA | AK040441.1 | - | 5 | 2934 | ||
BC026653.1 | - | 1 | 435 | |||
BC029250.1 | - | 3 | 1253 | |||
BC030871.1 | - | 4 | 2079 | |||
BC036158.1 | - | 1 | 272 | |||
BC039286.1 | - | 6 | 2656 | |||
BC069956.1 | - | 1 | 238 | |||
BC092067.1 | - | 1 | 309 |
