siRNA Details
Gene Symbol: Tle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083927.1 | NP_001077396.1 | 20 | 3354 | ||
NM_001083928.1 | NP_001077397.1 | 20 | 3300 | |||
NM_009389.2 | NP_033415.2 | 20 | 3324 | |||
GenBank mRNA | AK031322.1 | - | 18 | 2131 | ||
AK122524.1 | - | 20 | 2808 | |||
AK147635.1 | - | 11 | 7452 | |||
AK158785.1 | - | 19 | 2635 | |||
AK189910.1 | - | 3 | 277 | |||
AK198921.1 | - | 4 | 311 | |||
BC006672.1 | - | 9 | 1276 | |||
BC056465.1 | - | 20 | 2335 | 5709252 | ||
X73360.1 | - | 19 | 2507 |
