siRNA Details
Gene Symbol: Tle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083927.1 | NP_001077396.1 | 19 | 3253 | ||
NM_001083928.1 | NP_001077397.1 | 19 | 3199 | |||
NM_009389.2 | NP_033415.2 | 19 | 3223 | |||
GenBank mRNA | AK031322.1 | - | 17 | 2030 | ||
AK122524.1 | - | 19 | 2707 | |||
AK147635.1 | - | 10 | 7351 | |||
AK158785.1 | - | 18 | 2534 | |||
AK189910.1 | - | 2 | 176 | |||
AK198921.1 | - | 3 | 210 | |||
BC006672.1 | - | 8 | 1175 | |||
BC056465.1 | - | 19 | 2234 | 5709252 | ||
X73360.1 | - | 18 | 2406 |
