siRNA Details
Gene Symbol: Tle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083927.1 | NP_001077396.1 | 16 | 2905 | ||
NM_001083928.1 | NP_001077397.1 | 16 | 2851 | |||
NM_009389.2 | NP_033415.2 | 16 | 2875 | |||
GenBank mRNA | AK031322.1 | - | 14 | 1682 | ||
AK122524.1 | - | 16 | 2359 | |||
AK147635.1 | - | 7 | 7003 | |||
AK158785.1 | - | 15 | 2186 | |||
BC006672.1 | - | 5 | 827 | |||
BC056465.1 | - | 16 | 1886 | 5709252 | ||
X73360.1 | - | 15 | 2058 |
