siRNA Details
Gene Symbol: Tpst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009419.3 | NP_033445.2 | 4 | 1077 | ||
GenBank mRNA | AF049890.1 | - | 3 | 1023 | ||
AK036436.1 | - | 3 | 1006 | |||
AK075672.1 | - | 4 | 1077 | |||
AK149502.1 | - | 4 | 1042 | |||
AK163461.1 | - | 4 | 1041 | |||
BC003721.1 | - | 4 | 1068 | 3494892 |
