siRNA Details
Gene Symbol: Tpst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009419.3 | NP_033445.2 | 3 | 695 | ||
GenBank mRNA | AF049890.1 | - | 2 | 641 | ||
AK036436.1 | - | 2 | 624 | |||
AK075672.1 | - | 3 | 695 | |||
AK149379.1 | - | 3 | 699 | |||
AK149502.1 | - | 3 | 658 | |||
AK150082.1 | - | 3 | 655 | |||
AK163461.1 | - | 3 | 659 | |||
BC003721.1 | - | 3 | 686 | 3494892 |
