siRNA Details
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009428.2 | NP_033454.1 | 4 | 1576 | ||
GenBank mRNA | AF029983.1 | - | 4 | 1231 | ||
AF060107.1 | - | 4 | 1575 | |||
AJ006204.1 | - | 3 | 1173 | |||
AK038531.1 | - | 4 | 1400 | |||
AK047867.1 | - | 2 | 597 | |||
BC112972.1 | - | 4 | 1394 | 40049027 |
