siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF402611.1 | - | 7, 8 | 1328 | ||
AF402612.1 | - | 6, 7 | 1225 | |||
AF402613.1 | - | 7, 8 | 1375 | |||
AK028302.1 | - | 4, 5 | 777 | |||
AK034504.1 | - | 6, 7 | 1318 | |||
AK054242.1 | - | 7, 8 | 1316 | |||
AK134961.1 | - | 7, 8 | 1391 | |||
AK149626.1 | - | 7, 8 | 1379 | |||
BC129902.1 | - | 7, 8 | 1415 | 40111637 | ||
BC129903.1 | - | 7, 8 | 1416 | 40111639 | ||
BC145523.1 | - | 7, 8 | 1469 |
